Assay Details
Gene Family ID: | MIPF0000403, MIR482 |
Mature miRNA Sequence: | AUGGUCGAAGUAGGCAAAAUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Arabidopsis | ath-miR472-5p | MIMAT0032014 |
Arabidopsis lyrata | aly-miR472-5p | MIMAT0017589 |
Stem-loop Details
Arabidopsis
Arabidopsis lyrata
