Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAGUUGUUCGUGGUGGAUUCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-382 | MIMAT0017956 |
Stem-loop Details
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ssc-miR-382 |
|---|---|
| miRBase Accession Number: | MI0015911 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGUUGUUCGUGGUGGAUUCG |
| Species: | Pig |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAGUUGUUCGUGGUGGAUUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-382 | MIMAT0017956 |