Assay Details
Gene Family ID: | MIPF0000621, mir-1251 |
Mature miRNA Sequence: | CGCUUUGCUCAGCCAGUGUAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1251-3p | MIMAT0026741 |
| Mouse | mmu-miR-1251-3p | MIMAT0014825 |
| Dasypus novemcinctus | dno-miR-1251-3p | MIMAT0048034 |
| Guinea pig | cpo-miR-1251-3p | MIMAT0047408 |
| Rabbit | ocu-miR-1251-3p | MIMAT0048588 |
| Rhesus monkey | mml-miR-1251-3p | MIMAT0028243 |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey

Genomic Map