Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 12 - 97492301 - 97492301 [+] on 97492301 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1251-3p | MIMAT0026741 |
Human | hsa-miR-1251-5p | MIMAT0005903 |
Stem-loop Details
Human
