Assay Details
Gene Family ID: | MIPF0000621, mir-1251 |
Mature miRNA Sequence: | CGCUUUGCUCAGCCAGUGUAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1251-3p | MIMAT0026741 |
Mouse | mmu-miR-1251-3p | MIMAT0014825 |
Dasypus novemcinctus | dno-miR-1251-3p | MIMAT0048034 |
Guinea pig | cpo-miR-1251-3p | MIMAT0047408 |
Rabbit | ocu-miR-1251-3p | MIMAT0048588 |
Rhesus monkey | mml-miR-1251-3p | MIMAT0028243 |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey
