Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 10 - 92137392 - 92137392 [+] on 92137392 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1251-5p | MIMAT0014824 |
Mouse | mmu-miR-1251-3p | MIMAT0014825 |
Stem-loop Details
Mouse
