Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AACGGCAAUGACUUUUGUACCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548al | MIMAT0019024 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-548al |
|---|---|
| miRBase Accession Number: | MI0016851 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACGGCAAUGACUUUUGUACCA |
| Species: | Human |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AACGGCAAUGACUUUUGUACCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548al | MIMAT0019024 |