Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AUAUACAAGGGCAAGCUCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pteropus alecto | pal-miR-381-3p | MIMAT0040155 |
| Sheep | oar-miR-381-3p | MIMAT0019293 |
Stem-loop Details
Pteropus alecto
Sheep

Genomic Map