Assay Details
Gene Family ID: | MIPF0001008, mir-2765 |
Mature miRNA Sequence: | CAGCGGUCGAGGAGUUCCUUAAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Tribolium castaneum | tca-miR-2765-3p | MIMAT0018631 |
Stem-loop Details
Tribolium castaneum
