Assay Details
Gene Family ID: | MIPF0000812, mir-2188 |
Mature miRNA Sequence: | GAUGCGUGUGGUCAGACCUAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Anolis carolinensis | aca-miR-2188-3p | MIMAT0021883 |
Stem-loop Details
Anolis carolinensis

More Information
Related Products
miRBase Alias:
Aeromonas caviae Ae398: aca-miR-2188* (v18)
