Assay Details
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | CGGGGCAGCUCAGUACAAGACG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-486-3p | MIMAT0023968 |
Stem-loop Details
Cricetulus griseus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cgr-miR-486-3p |
|---|---|
| miRBase Accession Number: | MI0020530 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCAGCUCAGUACAAGACG |
| Species: | Cricetulus griseus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | CGGGGCAGCUCAGUACAAGACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-486-3p | MIMAT0023968 |