Assay Details
Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Gorilla gorilla | ggo-miR-1278 | MIMAT0024283 |
Stem-loop Details
Gorilla gorilla

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ggo-miR-1278 |
---|---|
miRBase Accession Number: | MI0020831 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUA |
Species: | Gorilla gorilla |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Gorilla gorilla | ggo-miR-1278 | MIMAT0024283 |