Assay Details
Gene Family ID: | MIPF0001464, mir-2114 |
Mature miRNA Sequence: | CGAGCCUCAAGCAAGGGACUUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pan troglodytes | ptr-miR-2114 | MIMAT0024072 |
Stem-loop Details
Pan troglodytes

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ptr-miR-2114 |
---|---|
miRBase Accession Number: | MI0020619 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGAGCCUCAAGCAAGGGACUUCA |
Species: | Pan troglodytes |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001464, mir-2114 |
Mature miRNA Sequence: | CGAGCCUCAAGCAAGGGACUUCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pan troglodytes | ptr-miR-2114 | MIMAT0024072 |