Assay Details
Gene Family ID: | MIPF0001437, mir-2763 |
Mature miRNA Sequence: | AUAUUAUGCUCAUUACUUUGGAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Manduca sexta | mse-miR-2763 | MIMAT0024489 |
Stem-loop Details
Manduca sexta

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mse-miR-2763 |
---|---|
miRBase Accession Number: | MI0021039 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AUAUUAUGCUCAUUACUUUGGAU |
Species: | Manduca sexta |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001437, mir-2763 |
Mature miRNA Sequence: | AUAUUAUGCUCAUUACUUUGGAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Manduca sexta | mse-miR-2763 | MIMAT0024489 |