Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pig | ssc-miR-489 | MIMAT0025375 |
Stem-loop Details
Pig

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ssc-miR-489 |
---|---|
miRBase Accession Number: | MI0022150 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
Species: | Pig |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pig | ssc-miR-489 | MIMAT0025375 |