Assay Details
Gene Family ID: | MIPF0001809, MIR4376 |
Mature miRNA Sequence: | UACGCAGGAGAGAUGAUGCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Solanum tuberosum | stu-miR391-5p | MIMAT0031274 |
Stem-loop Details
Solanum tuberosum
More Information
Related Products
miRBase Alias:
Siccibacter turicensis: stu-miR4376-5p (v21)

Genomic Map