Assay Details
Mature miRNA Sequence: | UCCCUGAGUAGCUGCCGUUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis briggsae | cbr-miR-2231 | MIMAT0029917 |
Stem-loop Details
Caenorhabditis briggsae
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cbr-miR-2231 |
|---|---|
| miRBase Accession Number: | MI0025047 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCUGAGUAGCUGCCGUUGA |
| Species: | Caenorhabditis briggsae |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCCCUGAGUAGCUGCCGUUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis briggsae | cbr-miR-2231 | MIMAT0029917 |