Assay Details
Mature miRNA Sequence: | AAUCAUUCACGGACAACACUUUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dasypus novemcinctus | dno-miR-382-3p | MIMAT0047867 |
Guinea pig | cpo-miR-382-3p | MIMAT0047228 |
Rabbit | ocu-miR-382-3p | MIMAT0048428 |
Rhesus monkey | mml-miR-382-3p | MIMAT0026868 |
Stem-loop Details
Dasypus novemcinctus
Guinea pig
Rabbit
Rhesus monkey
