Assay Details
Gene Family ID: | MIPF0001711, mir-1545 |
Mature miRNA Sequence: | AGUGCGCAGGGAUAGAUAAGAGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-1545a-5p | MIMAT0026769 |
Stem-loop Details
Monodelphis domestica

Genomic Map