Assay Details
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Eptesicus fuscus | efu-miR-486 | MIMAT0035060 |
| Pan paniscus | ppa-miR-486-2 | MIMAT0049066 |
| Saimiri boliviensis | sbo-miR-486b | MIMAT0049473 |
Stem-loop Details
Eptesicus fuscus
Pan paniscus
Saimiri boliviensis

Genomic Map