Assay Details
Gene Family ID: | MIPF0000010, MIR159 |
Mature miRNA Sequence: | AGCUGCCGAUUCAUUCAUUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Aegilops tauschii | ata-miR319-5p | MIMAT0037244 |
Stem-loop Details
Aegilops tauschii

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ata-miR319-5p |
---|---|
miRBase Accession Number: | MI0031725 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGCUGCCGAUUCAUUCAUUCA |
Species: | Aegilops tauschii |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000010, MIR159 |
Mature miRNA Sequence: | AGCUGCCGAUUCAUUCAUUCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Aegilops tauschii | ata-miR319-5p | MIMAT0037244 |