Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. 7 - 64205504 - 64205504 [+] on 64205504 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-211-3p | MIMAT0017059 |
Mouse | mmu-miR-211-5p | MIMAT0000668 |
Stem-loop Details
Mouse
