Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 9 - 122682675 - 122682675 [+] on 122682675 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-138-1-3p | MIMAT0004668 |
| Mouse | mmu-miR-138-5p | MIMAT0000150 |
Stem-loop Details
Mouse

Genomic Map