Assay Details
Amplicon Length: | 102 |
Chromosome Location: | Chr. 5 - 9375501 - 9375501 [+] on 9375501 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-879-3p | MIMAT0004843 |
| Mouse | mmu-miR-879-5p | MIMAT0004842 |
Stem-loop Details
Mouse

Genomic Map