Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. X - 103572718 - 103572718 [+] on 103572718 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-421-3p | MIMAT0004869 |
| Mouse | mmu-miR-421-5p | MIMAT0017273 |
Stem-loop Details
Mouse

Genomic Map