Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. X - 68010607 - 68010607 [+] on 68010607 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-509-3p | MIMAT0004891 |
| Mouse | mmu-miR-509-5p | MIMAT0004890 |
Stem-loop Details
Mouse

Genomic Map