Assay Details
Amplicon Length: | 103 |
Chromosome Location: | Chr. 15 - 35660675 - 35660675 [+] on 35660675 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-875-5p | MIMAT0004937 |
Mouse | mmu-miR-875-3p | MIMAT0004938 |
Stem-loop Details
Mouse
