Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 4 - 48213366 - 48213366 [+] on 48213366 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1958 | MIMAT0009431 |
Stem-loop Details
Mouse
