Assay Details
Amplicon Length: | 125 |
Chromosome Location: | Chr. 5 - 30170596 - 30170596 [-] on 30170596 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-1960 | MIMAT0009433 |
Stem-loop Details
Mouse
