Assay Details
Gene Family ID: | MIPF0000537, mir-572 |
Mature miRNA Sequence: | GUCCGCUCGGCGGUGGCCCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-572 | MIMAT0003237 |
| Pan troglodytes | ptr-miR-572 | MIMAT0008248 |
| Rhesus monkey | mml-miR-572 | MIMAT0006434 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey

Genomic Map