Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAAUGGUUCCCUUUAGAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-522-3p | MIMAT0002868 |
| Gorilla gorilla | ggo-miR-522-3p | MIMAT0036438 |
| Pan troglodytes | ptr-miR-522 | MIMAT0008206 |
| Pongo pygmaeus | ppy-miR-522 | MIMAT0015978 |
| Rhesus monkey | mml-miR-518e | MIMAT0006382 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-522 (v17)
Macaca mulatta: mml-miR-522 (v15)
Macaca mulatta: mml-miR-522 (v15)

Genomic Map