Assay Details
Gene Family ID: | MIPF0000613, mir-1202 |
Mature miRNA Sequence: | GUGCCAGCUGCAGUGGGGGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1202 | MIMAT0005865 |
Pan troglodytes | ptr-miR-1202 | MIMAT0007956 |
Pongo pygmaeus | ppy-miR-1202 | MIMAT0016152 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
