Assay Details
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCUUCUUUACAGUGUUGCCUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-107-5p | MIMAT0017048 |
| Chicken | gga-miR-107-5p | MIMAT0026520 |
| Columba livia | cli-miR-107-5p | MIMAT0038489 |
| Dasypus novemcinctus | dno-miR-107-5p | MIMAT0047608 |
| Guinea pig | cpo-miR-107-5p | MIMAT0046959 |
| Pteropus alecto | pal-miR-107a-5p | MIMAT0040201 |
| View More Rows | ||
| Python bivittatus | pbv-miR-107-5p | MIMAT0038907 |
| Rabbit | ocu-miR-107-5p | MIMAT0048199 |
| Rhesus monkey | mml-miR-107-5p | MIMAT0026584 |
| Salmo salar | ssa-miR-107-5p | MIMAT0032290 |
| Xenopus laevis | xla-miR-107-5p | MIMAT0046420 |
Stem-loop Details
Mouse
Chicken
Columba livia
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Python bivittatus
Rabbit
Rhesus monkey
Salmo salar
Xenopus laevis

Genomic Map