Assay Details
Amplicon Length: | 119 |
Chromosome Location: | Chr. 10 - 65291029 - 65291029 [+] on 65291029 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-451-3p | MIMAT0017193 |
| Rat | rno-miR-451-5p | MIMAT0001633 |
Stem-loop Details
Rat

Genomic Map