Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAGGUUGCCCUUGGUGAAUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-377-5p | MIMAT0004689 |
| Mouse | mmu-miR-377-5p | MIMAT0017079 |
| Rat | rno-miR-377-5p | MIMAT0017203 |
| Dog | cfa-miR-377 | MIMAT0009890 |
| Guinea pig | cpo-miR-377-5p | MIMAT0047217 |
| Rabbit | ocu-miR-377-5p | MIMAT0048419 |
| View More Rows | ||
| Rhesus monkey | mml-miR-377-5p | MIMAT0026864 |
Stem-loop Details
Human
Mouse
Rat
Dog
Guinea pig
Rabbit
Rhesus monkey

Genomic Map