Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAGGUUGCCCUUGGUGAAUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-377-5p | MIMAT0004689 | |
| Mouse | mmu-miR-377-5p | MIMAT0017079 | |
| Rat | rno-miR-377-5p | MIMAT0017203 | |
| Dog | cfa-miR-377 | MIMAT0009890 | |
| Guinea pig | cpo-miR-377-5p | MIMAT0047217 | |
| Rabbit | ocu-miR-377-5p | MIMAT0048419 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-377-5p | MIMAT0026864 | |
Stem-loop Details
Human
Mouse
Rat
Dog
Guinea pig
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-377* (v17)
Mouse: mmu-miR-377* (v17)
Rat: rno-miR-377* (v18)
Mouse: mmu-miR-377* (v17)
Rat: rno-miR-377* (v18)

Genomic Map