Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 18 - 16390353 - 16390353 [+] on 16390353 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-187-3p | MIMAT0000864 |
| Rat | rno-miR-187-5p | MIMAT0017144 |
Stem-loop Details
Rat

Genomic Map