Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | UCGUGUCUUGUGUUGCAGCCGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-187-3p | MIMAT0000262 |
Mouse | mmu-miR-187-3p | MIMAT0000216 |
Rat | rno-miR-187-3p | MIMAT0000864 |
Bovine | bta-miR-187 | MIMAT0009248 |
Callithrix jacchus | cja-miR-187 | MIMAT0039471 |
Dasypus novemcinctus | dno-miR-187-3p | MIMAT0047710 |
View More Rows | ||
Dog | cfa-miR-187 | MIMAT0009843 |
Guinea pig | cpo-miR-187-3p | MIMAT0047059 |
Horse | eca-miR-187 | MIMAT0012999 |
Pig | ssc-miR-187 | MIMAT0020587 |
Pteropus alecto | pal-miR-187-3p | MIMAT0040372 |
Rabbit | ocu-miR-187-3p | MIMAT0048296 |
Rhesus monkey | mml-miR-187-3p | MIMAT0006221 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pig
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-187 (v17)
Mouse: mmu-miR-187 (v17)
Rat: rno-miR-187 (v18)
Macaca mulatta: mml-miR-187 (v19)
Mouse: mmu-miR-187 (v17)
Rat: rno-miR-187 (v18)
Macaca mulatta: mml-miR-187 (v19)
