Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | UCGUGUCUUGUGUUGCAGCCGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-187-3p | MIMAT0000262 |
| Mouse | mmu-miR-187-3p | MIMAT0000216 |
| Rat | rno-miR-187-3p | MIMAT0000864 |
| Bovine | bta-miR-187 | MIMAT0009248 |
| Callithrix jacchus | cja-miR-187 | MIMAT0039471 |
| Dasypus novemcinctus | dno-miR-187-3p | MIMAT0047710 |
| View More Rows | ||
| Dog | cfa-miR-187 | MIMAT0009843 |
| Guinea pig | cpo-miR-187-3p | MIMAT0047059 |
| Horse | eca-miR-187 | MIMAT0012999 |
| Pig | ssc-miR-187 | MIMAT0020587 |
| Pteropus alecto | pal-miR-187-3p | MIMAT0040372 |
| Rabbit | ocu-miR-187-3p | MIMAT0048296 |
| Rhesus monkey | mml-miR-187-3p | MIMAT0006221 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pig
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map