Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | UCGUGUCUUGUGUUGCAGCCGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-187-3p | MIMAT0000262 | |
| Mouse | mmu-miR-187-3p | MIMAT0000216 | |
| Rat | rno-miR-187-3p | MIMAT0000864 | |
| Bovine | bta-miR-187 | MIMAT0009248 | |
| Callithrix jacchus | cja-miR-187 | MIMAT0039471 | |
| Dasypus novemcinctus | dno-miR-187-3p | MIMAT0047710 | |
| View More Rows | |||
| Dog | cfa-miR-187 | MIMAT0009843 | |
| Guinea pig | cpo-miR-187-3p | MIMAT0047059 | |
| Horse | eca-miR-187 | MIMAT0012999 | |
| Pig | ssc-miR-187 | MIMAT0020587 | |
| Pteropus alecto | pal-miR-187-3p | MIMAT0040372 | |
| Rabbit | ocu-miR-187-3p | MIMAT0048296 | |
| Rhesus monkey | mml-miR-187-3p | MIMAT0006221 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Pig
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-187 (v17)
Mouse: mmu-miR-187 (v17)
Rat: rno-miR-187 (v18)
Macaca mulatta: mml-miR-187 (v19)
Mouse: mmu-miR-187 (v17)
Rat: rno-miR-187 (v18)
Macaca mulatta: mml-miR-187 (v19)

Genomic Map