Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 7 - 74423084 - 74423084 [+] on 74423084 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-875 | MIMAT0012842 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Amplicon Length: | 67 |
Chromosome Location: | Chr. 7 - 74423084 - 74423084 [+] on 74423084 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-875 | MIMAT0012842 |