Assay Details
Gene Family ID: | MIPF0000084, mir-142 |
Mature miRNA Sequence: | CAUAAAGUAGAAAGCACUACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-142-5p | MIMAT0000433 |
| Mouse | mmu-miR-142a-5p | MIMAT0000154 |
| Rat | rno-miR-142-5p | MIMAT0000847 |
| Astatotilapia burtoni | abu-miR-142-5p | MIMAT0042103 |
| Columba livia | cli-miR-142-5p | MIMAT0038529 |
| Dasypus novemcinctus | dno-miR-142-5p | MIMAT0047657 |
| View More Rows | ||
| Fugu rubripes | fru-miR-142 | MIMAT0002908 |
| Gadus morhua | gmo-miR-142-5p | MIMAT0044045 |
| Guinea pig | cpo-miR-142-5p | MIMAT0047008 |
| Horse | eca-miR-142-5p | MIMAT0013022 |
| Metriaclima zebra | mze-miR-142 | MIMAT0042338 |
| Neolamprologus brichardi | nbr-miR-142 | MIMAT0042531 |
| Oreochromis niloticus | oni-miR-142 | MIMAT0042701 |
| Ornithorhynchus anatinus | oan-miR-142-5p | MIMAT0006982 |
| Pig | ssc-miR-142-5p | MIMAT0013919 |
| Pongo pygmaeus | ppy-miR-142-5p | MIMAT0015765 |
| Pundamilia nyererei | pny-miR-142 | MIMAT0042917 |
| Rabbit | ocu-miR-142-5p | MIMAT0048245 |
| Rhesus monkey | mml-miR-142-5p | MIMAT0006199 |
| Salmo salar | ssa-miR-142a-5p | MIMAT0032361 |
| Tetraodon nigroviridis | tni-miR-142a | MIMAT0002909 |
| Zebrafish | dre-miR-142a-5p | MIMAT0001838 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-142 |
| Stem-loop Accession # | MI0000458 |
| Stem-loop Sequence | GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG |
| Chromosome Location | Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000433
miRBase ID: hsa-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: Chr. 17 - 58331232 - 58331318 [-] on Build GRCh38 |
| Stem-loop ID | mmu-mir-142a |
| Stem-loop Accession # | MI0000167 |
| Stem-loop Sequence | ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000154
miRBase ID: mmu-miR-142a-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: Chr. 11 - 87756864 - 87756927 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-142 |
| Stem-loop Accession # | MI0000915 |
| Stem-loop Sequence | GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG |
| Chromosome Location | Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000847
miRBase ID: rno-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: Chr. 10 - 75050839 - 75050925 [+] on Build Rnor_6.0 |
| Stem-loop ID | abu-mir-142-1 |
| Stem-loop Accession # | MI0033805 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042103
miRBase ID: abu-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | abu-mir-142-2 |
| Stem-loop Accession # | MI0033806 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042103
miRBase ID: abu-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | cli-mir-142 |
| Stem-loop Accession # | MI0029977 |
| Stem-loop Sequence | GAGACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAAACAGCACUGCAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038529
miRBase ID: cli-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | dno-mir-142 |
| Stem-loop Accession # | MI0038991 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047657
miRBase ID: dno-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | fru-mir-142 |
| Stem-loop Accession # | MI0003223 |
| Stem-loop Sequence | ACAGUGCUGUCACCCAUAAAGUAGAAAGCACUACUAAACUAUUCUACACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGCACUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002908
miRBase ID: fru-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-142-1 |
| Stem-loop Accession # | MI0036183 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCCACAGUGUAGUGUUUCCUACUUUAUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044045
miRBase ID: gmo-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-142-2 |
| Stem-loop Accession # | MI0036000 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUCAAUGCAAAGUGUAGUGUUUCCUACUUUAUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044045
miRBase ID: gmo-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-142 |
| Stem-loop Accession # | MI0038649 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047008
miRBase ID: cpo-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | eca-mir-142 |
| Stem-loop Accession # | MI0012771 |
| Stem-loop Sequence | ACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013022
miRBase ID: eca-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | mze-mir-142-1 |
| Stem-loop Accession # | MI0034109 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042338
miRBase ID: mze-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | mze-mir-142-2 |
| Stem-loop Accession # | MI0034111 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042338
miRBase ID: mze-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | nbr-mir-142-1 |
| Stem-loop Accession # | MI0034376 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042531
miRBase ID: nbr-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | nbr-mir-142-2 |
| Stem-loop Accession # | MI0034377 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042531
miRBase ID: nbr-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | oni-mir-142-1 |
| Stem-loop Accession # | MI0034610 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042701
miRBase ID: oni-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | oni-mir-142-2 |
| Stem-loop Accession # | MI0034611 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042701
miRBase ID: oni-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | oan-mir-142 |
| Stem-loop Accession # | MI0006772 |
| Stem-loop Sequence | CGACAGACAGACAGACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAACAGCGCUGUAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUGGGCUUCGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006982
miRBase ID: oan-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-142 |
| Stem-loop Accession # | MI0013134 |
| Stem-loop Sequence | GUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013919
miRBase ID: ssc-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-142 |
| Stem-loop Accession # | MI0014833 |
| Stem-loop Sequence | GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015765
miRBase ID: ppy-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | pny-mir-142-1 |
| Stem-loop Accession # | MI0034893 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042917
miRBase ID: pny-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | pny-mir-142-2 |
| Stem-loop Accession # | MI0034895 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUUUACUACACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042917
miRBase ID: pny-miR-142 Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-142 |
| Stem-loop Accession # | MI0039300 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048245
miRBase ID: ocu-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | mml-mir-142 |
| Stem-loop Accession # | MI0007631 |
| Stem-loop Sequence | GACAGUGCAGUCACCCAUAAAGUAGAAAGCACUACUAACAGCACUGGAGGGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006199
miRBase ID: mml-miR-142-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-142a-1 |
| Stem-loop Accession # | MI0026500 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCACCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032361
miRBase ID: ssa-miR-142a-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | ssa-mir-142a-2 |
| Stem-loop Accession # | MI0026501 |
| Stem-loop Sequence | CAUAAAGUAGAAAGCACUACUAAACUCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032361
miRBase ID: ssa-miR-142a-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | tni-mir-142a |
| Stem-loop Accession # | MI0003224 |
| Stem-loop Sequence | GUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACUCUUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002909
miRBase ID: tni-miR-142a Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
| Stem-loop ID | dre-mir-142a |
| Stem-loop Accession # | MI0002005 |
| Stem-loop Sequence | CGUACAGUGCAGUCAUCCAUAAAGUAGAAAGCACUACUAAACCCCUCGCCACAGUGUAGUGUUUCCUACUUUAUGGAUGAGUGUACUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001838
miRBase ID: dre-miR-142a-5p Mature miRNA Sequence: CAUAAAGUAGAAAGCACUACU Chromosome Location: NA |
More Information
Related Products

Genomic Map