Assay Details
Gene Family ID: | MIPF0000353, mir-802 |
Mature miRNA Sequence: | ACGGAGAGUCUUUGUCACUCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-802-3p | MIMAT0017240 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-802-3p |
---|---|
Stem-loop Accession Number: | MI0004249 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACGGAGAGUCUUUGUCACUCAG |
Species: | Mouse |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000353, mir-802 |
Mature miRNA Sequence: | ACGGAGAGUCUUUGUCACUCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-802-3p | MIMAT0017240 |