Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUGGUCCGUGGCGCGUUCGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-323a-5p | MIMAT0004696 |
| Mouse | mmu-miR-323-5p | MIMAT0004638 |
| Rat | rno-miR-323-5p | MIMAT0004637 |
| Horse | eca-miR-323-5p | MIMAT0013131 |
| Pongo pygmaeus | ppy-miR-323-5p | MIMAT0015825 |
| Rabbit | ocu-miR-323a-5p | MIMAT0048368 |
| View More Rows | ||
| Rhesus monkey | mml-miR-323a-5p | MIMAT0006264 |
Stem-loop Details
Human
Mouse
Rat
Horse
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map