siRNA Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178729.4 | NP_848844.1 | 9 | 2065 | ||
| GenBank mRNA | AK029957.1 | - | 9 | 2013 | ||
| AK041497.1 | - | 9 | 2037 | |||
| AK082931.1 | - | 9 | 2035 | |||
| AK171783.1 | - | 9 | 1925 | |||
| BC113798.1 | - | 9 | 1983 | 40062506 | ||

Fbxl5
Genomic Map