siRNA Details
| Gene Symbol: AW551984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199556.1 | NP_001186485.1 | 4, 5 | 492 | ||
| NM_178737.5 | NP_848852.1 | 4, 5 | 482 | |||
| GenBank mRNA | AK028282.1 | - | 4, 5 | 456 | ||
| AK049098.1 | - | 4, 5 | 483 | |||
| AK082321.1 | - | 4, 5 | 465 | |||
| BC061211.1 | - | 4, 5 | 394 | 30250574 | ||

AW551984
Genomic Map