siRNA Details
| Gene Symbol: Hps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005247.2 | NP_001005247.2 | 2, 3 | 617 | ||
| NM_001005248.2 | NP_001005248.2 | 2, 3 | 593 | |||
| NM_001167864.1 | NP_001161336.1 | 2, 3 | 617 | |||
| GenBank mRNA | AF534396.1 | - | 2, 3 | 414 | ||
| AF534397.1 | - | 2, 3 | 390 | |||
| AF534399.1 | - | 2, 3 | 414 | |||
| AK129265.1 | - | 2, 3 | 544 | |||
| AK141088.1 | - | 2, 3 | 415 | |||
| AK154612.1 | - | 2, 3 | 435 | |||
| AK157182.1 | - | 2, 3 | 384 | |||
| BC046443.1 | - | 2, 3 | 232 | |||
| BC082542.1 | - | 2, 3 | 617 | 6405811 | ||

Hps5
Genomic Map