siRNA Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178743.3 | NP_848858.2 | 17 | 2243 | ||
| GenBank mRNA | AK052202.1 | - | 12 | 2330 | ||
| AK078545.1 | - | 11 | 3799 | |||
| AK089182.1 | - | 17 | 2223 | |||
| BC132493.1 | - | 12 | 2111 | 40130770 | ||
| BC137897.1 | - | 12 | 2111 | 8860916 | ||

Slc26a11
Genomic Map