siRNA Details
| Gene Symbol: Slc25a40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289595.1 | NP_001276524.1 | 4 | 435 | ||
| NM_001289596.1 | NP_001276525.1 | 5 | 488 | |||
| NM_178766.5 | NP_848881.2 | 5 | 507 | |||
| GenBank mRNA | AK044260.1 | - | 508 | |||
| AK045863.1 | - | 5 | 495 | |||
| AK079538.1 | - | 5 | 437 | |||
| AK084797.1 | - | 4 | 375 | |||
| AK088444.1 | - | 5 | 473 | |||
| AK136770.1 | - | 5 | 469 | |||
| AK163037.1 | - | 6 | 564 | |||
| BC083103.1 | - | 4 | 285 | 30726859 | ||

Slc25a40
Genomic Map