siRNA Details
| Gene Symbol: Rnf152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160368.1 | NP_001153840.1 | 2 | 571 | ||
| NM_178779.4 | NP_848894.1 | 3 | 694 | |||
| GenBank mRNA | AK035832.1 | - | 3 | 490 | ||
| AK044638.1 | - | 3 | 655 | |||
| BC118961.2 | - | 2 | 317 | 40094497 | ||
| CJ112559.1 | - | 2 | 571 | |||

Rnf152
Genomic Map